1-Benzyl-4-piperidone 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12390-14 A12390-22 A12390-36
CAAAA12390-14EA 53.25 CAD
CAAAA12390-14 CAAAA12390-22 CAAAA12390-36
1-Benzyl-4-piperidone 98+%
1-Benzyl-4-piperidone

Purity: 98+% CAS No.: 3612-20-2 Molecular Formula: C12H15NO Formula Weight: 189.26 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5399 Boiling Point: 248° Beilstein Reference: 128556 MDL No.: MFCD00006192

Formula: C₁₂H₁₅NO
Boiling Pt: 248 °C
Density: 1.059
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006192
CAS Number: 3612-20-2
EINECS: 222-782-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR