Sodium chloride 99+%, crystalline powder PDV

Supplier: Thermo Scientific Chemicals

A12313-0B A12313-0G A12313-0I
CAAAAA12313-0BEA 62.09 CAD
CAAAAA12313-0B CAAAAA12313-0G CAAAAA12313-0I
Sodium chloride 99+%, crystalline powder PDV
Sodium chloride

Purity: 99+% CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Refractive Index: 1.5442 Odor: Odorless pH: 6-9 Melting Point: 801° Boiling Point: 1413° Merck Reference: 14,8599 MDL No.: MFCD00003477

Formula: NaCl
Boiling Pt: 1413 °C
Melting Pt: 801 °C
Density: 2.165
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,8599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR