Potassium oxalate hydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

A12289-0B A12289-0I A12289-30
CAAAAA12289-0BEA 169.06 CAD
CAAAAA12289-0B CAAAAA12289-0I CAAAAA12289-30
Potassium oxalate hydrate 99%
Potassium oxalate hydrate

Purity: 99% CAS No.: 6487-48-5 Molecular Formula: K2C2O4.xH2O Formula Weight: 166.22(anhy) Odor: Odorless pH: 6-8.5 Melting Point: 300° Beilstein Reference: 3752576 Merck Reference: 14,7651 MDL No.: MFCD00150033

Formula: (COOK)₂·xH₂O
Melting Pt: >300 °C
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00150033
CAS Number: 6487-48-5
EINECS: 209-506-8
ADR: 6.1,III
Merck Index: 14,7651

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR