Anthranilonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Cyanoaniline, o-Cyanoaniline, 2-Aminobenzonitrile, o-Aminobenzonitrile

A12286-18 A12286-30
CAAAA12286-18EA 84.21 CAD
CAAAA12286-18 CAAAA12286-30
Anthranilonitrile 98%
Anthranilonitrile

Purity: 98% CAS No.: 1885-29-6 Molecular Formula: C7H6N2 Formula Weight: 118.14 Flash Point: 145°(293°F) Odor: Almond-like Boiling Point: 286° Beilstein Reference: 907187 UN No.: 3439 MDL No.: MFCD00007631

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 286 °C
Melting Pt: 45…49 °C
Flash Pt: 145 °C (293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007631
CAS Number: 1885-29-6
EINECS: 217-549-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR