tri-Sodium citrate dihydrate 99%

Supplier: Thermo Scientific Chemicals

A12274-0C A12274-0E A12274-36
CAAAAA12274-0CEA 603.56 CAD
CAAAAA12274-0C CAAAAA12274-0E CAAAAA12274-36
tri-Sodium citrate dihydrate 99%
tri-Sodium citrate dihydrate

Purity: 99% CAS No.: 6132-04-3 Molecular Formula: Na3C6H5O7.2H2O Formula Weight: 294.10 (258.07anhy) Odor: Odorless Melting Point: 150° -2H2O Beilstein Reference: 6104939 Merck Reference: 14,8602 MDL No.: MFCD00150031

Formula: Na₃C₆H₅O₇·2H₂O
Melting Pt: 150 °C (–H₂O of crystall.)
Density: 1.76
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 14,8602
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR