15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12265-03
A12265-06
A12265-14
CAAAAA12265-03EA
38.87
CAD
CAAAAA12265-03
CAAAAA12265-06
CAAAAA12265-14
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%
15-Crown-5 (1,4,7,10,13-Pentaoxacyclopentadecane, Lead ionophore V)
Purity: 98%
CAS No.: 33100-27-5
Molecular Formula: C10H20O5
Formula Weight: 220.27
Flash Point:
Formula:
C₁₀H₂₀O₅ Boiling Pt: 150…155 °C/0.5 mm Melting Pt: –20 °C Density: 1,110 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005110 CAS Number: 33100-27-5 EINECS: 251-379-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...