1,3,5-Trichloro-2-nitrobenzene 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,4,6-Trichloronitrobenzene, 1-Nitro-2,4,6-trichlorobenzene

A12262-14 A12262-22
CAAAA12262-14EA 158 CAD
CAAAA12262-14 CAAAA12262-22
1,3,5-Trichloro-2-nitrobenzene 98+%
1,3,5-Trichloro-2-nitrobenzene

Purity: 98+% CAS No.: 18708-70-8 Molecular Formula: C6H2Cl3NO2 Formula Weight: 226.45 Melting Point: 68-73° Beilstein Reference: 2213282 UN No.: 2811 MDL No.: MFCD00014690

Formula: C₆H₂Cl₃NO₂
MW: 226.45 g/mol
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00014690
CAS Number: 18708-70-8
EINECS: 242-518-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR