N-Methyl-2-pyrrolidone (NMP) 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12260-0C
A12260-0E
A12260-36
CAAAAA12260-0CEA
587.76
CAD
CAAAAA12260-0C
CAAAAA12260-0E
CAAAAA12260-36
N-Methyl-2-pyrrolidone (NMP) 99+%
N-Methyl-2-pyrrolidone (NMP)
Purity: 99+%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Refractive Index: 1.4705
Odor: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Formula:
C₅H₉NO Boiling Pt: 204 °C Melting Pt: –24 °C Density: 1.033 Flash Pt: 86 °C(187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003193 CAS Number: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...