Salicylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Hydroxybenzoic acid, o-Hydroxybenzoic acid
A12253-0E
A12253-30
A12253-36
CAAAAA12253-0EEA
197.5
CAD
CAAAAA12253-0E
CAAAAA12253-30
CAAAAA12253-36
Salicylic acid 99%
Salicylic acid
Purity: 99%
CAS No.: 69-72-7
Molecular Formula: C7H6O3
Formula Weight: 138.12
Flash Point: 157°(315°F)
Melting Point: 158-160°
Boiling Point: 211°/20mm
Beilstein Reference: 774890
Merck Reference: 14,8332
MDL No.: MFCD00002439
Formula:
(HO)C₆H₄CO₂H MW: 138.12 g/mol Boiling Pt: 336 Melting Pt: 158…160 °C Density: 1.443 Flash Pt: 157 °C (315 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002439 CAS Number: 69-72-7 EINECS: 200-712-3 Merck Index: 14,8332 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...