4-Chlorobutyryl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A12249-09 A12249-0B A12249-30
CAAAAA12249-09EA 44.24 CAD
CAAAAA12249-09 CAAAA12249-0B CAAAA12249-30
4-Chlorobutyryl chloride 98%
4-Chlorobutyryl chloride

Purity: 98% CAS No.: 4635-59-0 Molecular Formula: C4H6Cl2O Formula Weight: 141.00 Flash Point: 85°(185°F) Refractive Index: 1.4610 Odor: Acrid Melting Point: -49° Boiling Point: 173-174° Beilstein Reference: 773860 UN No.: 2927 MDL No.: MFCD00000754

Formula: C₄H₆Cl₂O
Boiling Pt: 173…174 °C
Melting Pt: –49 °C
Density: 1.258
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000754
CAS Number: 4635-59-0
EINECS: 225-059-1
UN: 2922
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR