Sodium hydrogen selenite 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12173-18
A12173-30
CAAAAA12173-0BEA
2112.14
CAD
CAAAAA12173-0B
CAAAAA12173-18
CAAAAA12173-30
Sodium hydrogen selenite 96%
Sodium hydrogen selenite
Purity: 96%
CAS No.: 7782-82-3
Molecular Formula: HNaO3Se
Formula Weight: 150.96
UN No.: 2630
MDL No.: MFCD00003525
Formula:
HNaO₃Se Storage Temperature: Ambient |
MDL Number:
MFCD00003525 CAS Number: 7782-82-3 EINECS: 231-966-3 UN: 2630 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...