N-Methylethylenediamine 95% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12165-06
A12165-14
A12165-22
CAAAA12165-06EA
108.55
CAD
CAAAA12165-06
CAAAA12165-14
CAAAA12165-22
N-Methylethylenediamine 95% stabilized
N-Methylethylenediamine
Formula:
C₃H₁₀N₂ Boiling Pt: 114…116 °C Density: 0.851 Flash Pt: 41 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008165 CAS Number: 109-81-9 EINECS: 203-707-4 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...