4-Bromobutyryl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A12094-06 A12094-14 A12094-22
CAAAA12094-06EA 61.62 CAD
CAAAA12094-06 CAAAA12094-14 CAAAA12094-22
4-Bromobutyryl chloride 97%
4-Bromobutyryl chloride

Purity: 97% CAS No.: 927-58-2 Molecular Formula: C4H6BrClO Formula Weight: 185.45 Flash Point: 91°(195°F) Refractive Index: 1.4920 Boiling Point: 102-103°/40mm Beilstein Reference: 1743026 UN No.: 3265 MDL No.: MFCD00000753

Formula: C₄H₆BrClO
Boiling Pt: 102…103 °C/40 mm
Density: 1.600
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000753
CAS Number: 927-58-2
EINECS: 213-154-0
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR