Succinic acid 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Butanedioic acid

A12084-0C A12084-0E A12084-36
CAAAAA12084-0CEA 1036.48 CAD
CAAAAA12084-0C CAAAAA12084-0E CAAAAA12084-36
Succinic acid 99+%
Succinic acid

Purity: 99+% CAS No.: 110-15-6 Molecular Formula: HO2CCH2CH2CO2H Formula Weight: 118.09 Flash Point: 206°(402°F) Melting Point: 186-188° Boiling Point: 235° Beilstein Reference: 1754069 Merck Reference: 14,8869 MDL No.: MFCD00002789

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C
Melting Pt: 186…188 °C
Density: 1.56
Flash Pt: 206 °C(402 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
ADR: 8,III
Merck Index: 14,8869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR