Potassium hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

A12080-0B A12080-18 A12080-30
CAAAAA12080-0BEA 543.52 CAD
CAAAAA12080-0B CAAAAA12080-18 CAAAAA12080-30
Potassium hexafluorophosphate 98%
Potassium hexafluorophosphate

Purity: 98% CAS No.: 17084-13-8 Molecular Formula: KPF6 Formula Weight: 184.07 Odor: Odorless pH: 3.0-7.0 Melting Point: 575° UN No.: 3260 MDL No.: MFCD00011412

Formula: F₆KP
Melting Pt: 575 °C
Density: 2.55
Storage Temperature: Ambient
MDL Number: MFCD00011412
CAS Number: 17084-13-8
EINECS: 241-143-0
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR