Hexabutylditin 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12007-09
A12007-18
A12007-30
CAAAAA12007-09EA
84.21
CAD
CAAAAA12007-09
CAAAAA12007-18
CAAAAA12007-30
Hexabutylditin 98%
Hexabutylditin
Purity: 98%
CAS No.: 813-19-4
Molecular Formula: C24H54Sn2
Formula Weight: 580.07
Storage Temperature: -30°C to -10°C
Flash Point: 124°(255°F)
Refractive Index: 1.5120
Boiling Point: 197-198°/10mm
Beilstein Reference: 3605476
UN No.: 2788
MDL No.: MFCD00009417
Formula:
C₂₄H₅₄Sn₂ Boiling Pt: 197…198 °C/10 mm Density: 1.152 Flash Pt: 124 °C (255 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00009417 CAS Number: 813-19-4 EINECS: 212-383-3 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...