4-Ethoxybenzaldehyde 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11987-14
A11987-22
A11987-36
CAAAAA11987-14EA
59.72
CAD
CAAAAA11987-14
CAAAAA11987-22
CAAAAA11987-36
4-Ethoxybenzaldehyde 97+%
4-Ethoxybenzaldehyde
Purity: 97+%
CAS No.: 10031-82-0
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point:
Formula:
C₉H₁₀O₂ Boiling Pt: 253…255 °C Melting Pt: 13…14 °C Density: 1,08 Flash Pt: <110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003388 CAS Number: 10031-82-0 EINECS: 233-093-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...