2,6-Dichlorophenylhydrazinium chloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dichlorophenylhydrazine hydrochloride, 1-(2,6-Dichlorophenyl)hydrazine hydrochloride

A11968-03 A11968-06
CAAAA11968-03EA 53.25 CAD
CAAAA11968-03 CAAAA11968-06 CAAAA11968-14
2,6-Dichlorophenylhydrazinium chloride 98+%
2,6-Dichlorophenylhydrazinium chloride

Purity: 98+% CAS No.: 50709-36-9 Molecular Formula: C6H6Cl2N2.HCl Formula Weight: 213.50 Melting Point: ca 225° dec. Beilstein Reference: 4569738

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Melting Pt: ∼ 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012930
CAS Number: 50709-36-9
EINECS: 256-730-7
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR