Starch, soluble (from potato)

Supplier: Thermo Scientific Chemicals

Synonyms: StarchVitex® iodine indicatorAmylodextrin

A11961-36 A11961-36 A11961-0E
CAAAA11961-36EA 97 CAD
CAAAA11961-36 AAA11961-36 AAAA11961-0E
Starch, soluble (from potato)
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Melting Pt: 256…258 °C (dec.)
MDL Number: MFCD00082026
CAS Number: 9005-84-9
Merck Index: 14,8799

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR