Antimony(III) chloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

A11944-0E A11944-22 A11944-36
CAAAAA11944-0EEA 586.18 CAD
CAAAAA11944-0E CAAAAA11944-22 CAAAAA11944-36
Antimony(III) chloride 99+%
Antimony (III) chloride

Purity: 99+% CAS No.: 10025-91-9 Molecular Formula: SbCl3 Formula Weight: 228.11 Odor: Pungent Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Formula: SbCl₃
Boiling Pt: 223.5 °C
Melting Pt: 73.4 °C
Density: 3.14
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 14,707

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR