Nitrilotriacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A11936-0E A11936-22 A11936-36
CAAAAA11936-0EEA 524.56 CAD
CAAAAA11936-0E CAAAAA11936-22 CAAAAA11936-36
Nitrilotriacetic acid 99%
Nitrilotriacetic acid

Purity: 99% CAS No.: 139-13-9 Molecular Formula: C6H9NO6 Formula Weight: 191.14 Odor: Odorless Melting Point: 246° dec. Beilstein Reference: 1710776 Merck Reference: 14,6579 MDL No.: MFCD00004287

Formula: C₆H₉NO₆
Boiling Pt: 498 °C (1013 hPa)
Melting Pt: 246 °C (dec.)
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004287
CAS Number: 139-13-9
EINECS: 205-355-7
ADR: 8,II
Merck Index: 14,6579

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR