Methyl-3,4,5-trimethoxybenzoate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,4,5-Trimethoxybenzoic acid methyl ester, Methyl 3,4,5-trimethoxybenzoate, Trimethylgallic Acid Methyl Ester

A11933-14 A11933-22 A11933-36
CAAAA11933-14EA 34.44 CAD
CAAAA11933-14 CAAAA11933-22 CAAAA11933-36
Methyl-3,4,5-trimethoxybenzoate 98+%
Methyl 3,4,5-rimethoxybenzoate

Purity: 98+% CAS No.: 1916-07-0 Molecular Formula: C11H14O5 Formula Weight: 226.23 Melting Point: 81-85° Boiling Point: 274-275° MDL No.: MFCD00008431

Formula: C₁₁H₁₄O₅
MW: 226.23 g/mol
Boiling Pt: 274…275 °C
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008431
CAS Number: 1916-07-0
EINECS: 217-629-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR