cis-Stilbene 97%

Supplier: Thermo Scientific Chemicals

A11924-03 A11924-06 A11924-14
CAAAAA11924-03EA 52.3 CAD
CAAAAA11924-03 CAAAAA11924-06 CAAAAA11924-14
cis-Stilbene 97%
cis-Stilbene

Purity: 97% CAS No.: 645-49-8 Molecular Formula: C14H12 Formula Weight: 180.25 Flash Point: 110°(230°F) Refractive Index: 1.6220 Odor: Weak Melting Point: 1-2° Boiling Point: 307° Beilstein Reference: 1616739 Merck Reference: 14,8817 MDL No.: MFCD00004788

Formula: (C₆H₅)CHCH(C₆H₅)
Boiling Pt: 307 °C
Melting Pt: 1…2 °C
Density: 1.04
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004788
CAS Number: 645-49-8
EINECS: 211-445-7
Merck Index: 14,8817

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR