3-Bromo-o-toluidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11884-06 A11884-14
CAAAAA11884-22EA 1900.82 CAD
CAAAAA11884-22 CAAAA11884-06 CAAAA11884-14
3-Bromo-o-toluidine 98+%
3-Bromo-o-toluidine

Purity: 98+% CAS No.: 55289-36-6 Molecular Formula: C7H8BrN Formula Weight: 186.06 Flash Point: 110°(230°F) Refractive Index: 1.6160 Boiling Point: 253-255° Beilstein Reference: 2354213 UN No.: 2810 MDL No.: MFCD00051579

Formula: C₇H₈BrN
Boiling Pt: 253…255 °C
Density: 1.510
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051579
CAS Number: 55289-36-6
EINECS: 259-568-5
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR