1-Aminocyclohexanecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Amino-1-cyclohexanecarboxylic acid, 1-Aminocyclohexane-1-carboxylic acid

A11853-06 A11853-14
CAAAAA11853-06EA 54.67 CAD
CAAAAA11853-06 CAAAAA11853-14
1-Aminocyclohexanecarboxylic acid 98%
1-Aminocyclohexanecarboxylic acid

Purity: 98% CAS No.: 2756-85-6 Molecular Formula: C7H13NO2 Formula Weight: 143.19 Melting Point: 300° Beilstein Reference: 2355692

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Boiling Pt: 272
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001487
CAS Number: 2756-85-6
EINECS: 220-411-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR