2,2,3,3,3-Pentafluoro-1-propanol 98%

Supplier: Thermo Scientific Chemicals
Warning

A11846-06 A11846-14
CAAAAA11846-06EA 61.3 CAD
CAAAAA11846-06 CAAAAA11846-14
2,2,3,3,3-Pentafluoro-1-propanol 98%
2,2,3,3,3-Pentafluoro-1-propanol

Purity: 98% CAS No.: 422-05-9 Molecular Formula: CF3CF2CH2OH Formula Weight: 150.05 Refractive Index: 1.2880 Boiling Point: 80-81° MDL No.: MFCD00004673

Formula: C₃H₃F₅O
Boiling Pt: 80…81 °C
Density: 1.505
Storage Temperature: Ambient
MDL Number: MFCD00004673
CAS Number: 422-05-9
EINECS: 207-012-7
UN: 1986
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR