Chromium(III) potassium sulfate dodecahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11816-0E
A11816-22
A11816-36
CAAAAA11816-0EEA
353.92
CAD
CAAAAA11816-0E
CAAAAA11816-22
CAAAAA11816-36
Chromium(III) potassium sulfate dodecahydrate 98%
Chromium (III) potassium sulphate dodecahydrate
Purity: 98%
CAS No.: 7788-99-0
Molecular Formula: CrKO8S2.12H2O
Formula Weight: 499.39 (283.21anhy)
Odor: Odorless
Melting Point: 89°
MDL No.: MFCD00149917
Formula:
CrK(SO₄)₂·12H₂O Melting Pt: 89 °C Density: 1.83 Storage Temperature: Ambient |
MDL Number:
MFCD00149917 CAS Number: 7788-99-0 EINECS: 233-401-6 Merck Index: 14,2229 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...