Manganese(II) sulfate monohydrate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11809-0E
A11809-36
CAAAAA11809-0EEA
251.22
CAD
CAAAAA11809-0E
CAAAAA11809-36
Manganese(II) sulfate monohydrate 97%
Manganese (II) sulphate monohydrate
Purity: 97%
CAS No.: 10034-96-5
Molecular Formula: MnSO4.H2O
Formula Weight: 169.02 (151.00anhy)
Odor: Odorless
pH: 3-3.5
Merck Reference: 14,5739
UN No.: 3077
MDL No.: MFCD00149159
Formula:
MnSO₄·1H₂O Melting Pt: 700 °C Density: 2.95 Storage Temperature: Ambient |
MDL Number:
MFCD00149159 CAS Number: 10034-96-5 EINECS: 232-089-9 UN: 3077 ADR: 9,III Merck Index: 14,5739 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...