Sodium metaborate tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

A11803-0C A11803-0E A11803-36
CAAAAA11803-0CEA 673.08 CAD
CAAAAA11803-0C CAAAAA11803-0E CAAAAA11803-36
Sodium metaborate tetrahydrate 98%
Sodium metaborate tetrahydrate

Purity: 98% CAS No.: 10555-76-7 Molecular Formula: NaBO2.4H2O Formula Weight: 137.86 (65.80anhy) Odor: Odorless Melting Point: ca 53° dec. MDL No.: MFCD00149205

Formula: BNaO₂·4H₂O
MW: 137.86 g/mol
Melting Pt: ca 53 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149205
CAS Number: 10555-76-7
EINECS: 231-891-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR