Sodium octyl sulfate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Octyl sulphate, sodium salt, Octyl sulfate, sodium salt, Octyl sulfuric acid salt, Octyl sulphuric acid salt

A11786-03 A11786-06 A11786-14
CAAAAA11786-03EA 65.57 CAD
CAAAAA11786-03 CAAAAA11786-06 CAAAAA11786-14
Sodium octyl sulfate 99%
Sodium octyl sulphate

Purity: 99% CAS No.: 142-31-4 Molecular Formula: CH3(CH2)7OSO3Na Formula Weight: 232.28 Melting Point: ca 195° dec. Beilstein Reference: 3658635

Formula: C₈H₁₇NaO₄S
MW: 232.28 g/mol
Melting Pt: ∼ 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007470
CAS Number: 142-31-4
EINECS: 205-535-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR