Barium sulfate 99%, precipitated

Supplier: Thermo Scientific Chemicals

A11768-0C A11768-0E A11768-36
CAAAAA11768-0CEA 603.56 CAD
CAAAAA11768-0C CAAAAA11768-0E CAAAAA11768-36
Barium sulfate 99%, precipitated
Barium sulphate

Purity: 99% CAS No.: 7727-43-7 Molecular Formula: BaSO4 Formula Weight: 233.40 Odor: Odorless Melting Point: 1580° dec. MDL No.: MFCD00003455

Formula: BaSO₄
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 1580 °C (dec.)
Density: 4.5
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 14,994

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR