Bismuth(III) nitrate pentahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A11748-22 A11748-36
CAAAAA11748-22EA 70.31 CAD
CAAAAA11748-22 CAAAAA11748-36
Bismuth(III) nitrate pentahydrate 98%
Bismuth (III) nitrate pentahydrate

Purity: 98% CAS No.: 10035-06-0 Molecular Formula: Bi(NO3)3.5H2O Formula Weight: 485.07 (394.99anhy) Odor: Odorless Melting Point: 30° dec. UN No.: 1477 MDL No.: MFCD00149157

Formula: Bi(NO₃)₃·5H₂O
Boiling Pt: 75 - 80 °C (1013 hPa)
Melting Pt: 30 °C (dec.)
Density: 2.83
Storage Temperature: Ambient
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,III
Merck Index: 14,1271

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR