Lead(II) acetate trihydrate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11746-0B
A11746-30
CAAAAA11746-0BEA
180.12
CAD
CAAAAA11746-0B
CAAAAA11746-30
Lead(II) acetate trihydrate 99%
Lead (II) acetate trihydrate
Purity: 99%
CAS No.: 6080-56-4
Molecular Formula: Pb(OOCCH3)2.3H2O
Formula Weight: 379.33 (325.28anhy)
Melting Point: 75°
Beilstein Reference: 3730298
Merck Reference: 14,5397
UN No.: 1616
MDL No.: MFCD00150023
Formula:
(H₃CCOO)₂Pb·3H₂O Boiling Pt: 280 °C (1013 hPa) Melting Pt: 75 °C Density: 2.55 Storage Temperature: Ambient |
MDL Number:
MFCD00150023 CAS Number: 6080-56-4 EINECS: 206-104-4 UN: 1616 ADR: 6.1,III Merck Index: 14,5397 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...