2-Pyridinethiol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridine-2-thiol, 2-Mercaptopyridine

A11741-06 A11741-14 A11741-22
CAA11741-06EA 53.09 CAD
CAA11741-06 CAA11741-14 CAA11741-22
2-Pyridinethiol 98%
2-Pyridinethiol

Purity: 98% CAS No.: 2637-34-5 Molecular Formula: C5H5NS Formula Weight: 111.17 Storage Temperature: 2°C to 8°C Melting Point: 124-130° Beilstein Reference: 105758 MDL No.: MFCD00006285

Formula: C₅H₅NS
MW: 111.17 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 124…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00006285
CAS Number: 2637-34-5
EINECS: 220-131-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR