Benzyl-4-hydroxybenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzyl 4-hydroxybenzoate, Benzylparaben, 4-Hydroxybenzoic acid benzyl ester, Benzyl Parahydroxybenzoate

A11735-09 A11735-18
CAAAAA11735-09EA 53.88 CAD
CAAAAA11735-09 CAAAAA11735-18
Benzyl-4-hydroxybenzoate 99%
Benzyl 4-hydroxybenzoate

Purity: 99% CAS No.: 94-18-8 Molecular Formula: C14H12O3 Formula Weight: 228.25 Melting Point: 110-114° Beilstein Reference: 2115995

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00016471
CAS Number: 94-18-8
EINECS: 202-311-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR