4-(Dimethylamino)benzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde

A11712-0E A11712-22 A11712-36
CAAAAA11712-0EEA 1001.72 CAD
CAAAAA11712-0E CAAAAA11712-22 CAAAAA11712-36
4-(Dimethylamino)benzaldehyde 98%
4-(Dimethylamino)benzaldehyde

Purity: 98% CAS No.: 100-10-7 Molecular Formula: C9H11NO Formula Weight: 149.19 Flash Point: 164°(327°F) Odor: Strong Boiling Point: 176-177°/17mm Beilstein Reference: 606802 Merck Reference: 14,3230 MDL No.: MFCD00003381

Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176-177
Melting Pt: 72
Flash Pt: 164 °C(327 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
Merck Index: 14,3230
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR