4-Ethylbenzoyl chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

A11698-09 A11698-18
CAAAA11698-09EA 76.63 CAD
CAAAA11698-09 CAAAA11698-18
4-Ethylbenzoyl chloride 98+%
4-Ethylbenzoyl chloride

Purity: 98+% CAS No.: 16331-45-6 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Refractive Index: 1.5479 Odor: Acidic Boiling Point: 235-236° Beilstein Reference: 2206041 UN No.: 3265 MDL No.: MFCD00000697

Formula: C₉H₉ClO
Boiling Pt: 235…236 °C
Melting Pt: 42…43 °C
Density: 1.146
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000697
CAS Number: 16331-45-6
EINECS: 240-404-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR