Ethanolamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Aminoethanol, Monoethanolamine
A11697-0E
A11697-36
CAAAAA11697-0EEA
143.46
CAD
CAAAAA11697-0E
CAAAAA11697-30
CAAAAA11697-36
Ethanolamine 98+%
Ethanolamine
Purity: 98+%
CAS No.: 141-43-5
Molecular Formula: C2H7NO
Formula Weight: 61.08
Flash Point: 93°(199°F)
Refractive Index: 1.4540
Odor: Ammonia-like
Melting Point: 10°
Boiling Point: 169-170°
Beilstein Reference: 505944
Merck Reference: 14,3727
UN No.: 2491
MDL No.: MFCD00008183
Formula:
NH₂CH₂CH₂OH MW: 61.08 g/mol Boiling Pt: 171 Melting Pt: 10.5 Density: 1.012 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008183 CAS Number: 141-43-5 EINECS: 205-483-3 UN: 2491 ADR: 8 III Merck Index: 14,3727 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...