D-(+)-Malic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11688-14
A11688-06
CAAAAA11688-14EA
508.76
CAD
CAAAAA11688-14
CAAAA11688-03
CAAAA11688-06
D-(+)-Malic acid 98+%
D(+)-Malic acid
Purity: 98+%
CAS No.: 636-61-3
Molecular Formula: C4H6O5
Formula Weight: 134.09
Melting Point: 100-104°
Beilstein Reference: 1723540
Merck Reference: 14,5707
Optical Rotation: +28° (c=5.5 in pyridine)
MDL No.: MFCD00004245
Formula:
C₄H₆O₅ Boiling Pt: 322 °C (1013 hPa) Melting Pt: 100…104 °C Density: 1.60 Storage Temperature: Ambient |
MDL Number:
MFCD00004245 CAS Number: 636-61-3 EINECS: 211-262-2 Merck Index: 14,5707 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...