(±)-1,3-Butanediol 99%
Supplier: Thermo Scientific Chemicals
A11684-0D
A11684-0F
A11684-AP
CAAAAA11684-0DEA
410.8
CAD
CAAAAA11684-0D
CAAAAA11684-0F
CAAAAA11684-AP
(±)-1,3-Butanediol 99%
(±)-1,3-Butanediol
Purity: 99%
CAS No.: 107-88-0
Molecular Formula: C4H10O2
Formula Weight: 90.12
Flash Point: 109°(249°F)
Refractive Index: 1.4400
Odor: Mild
Melting Point: -50°
Boiling Point: 203-204°
Beilstein Reference: 1731276
Merck Reference: 14,1567
MDL No.: MFCD00004554
Formula:
C₄H₁₀O₂ Boiling Pt: 203…204 °C Melting Pt: –50 °C Density: 1.005 Flash Pt: 109 °C(249 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004554 CAS Number: 107-88-0 EINECS: 203-529-7 Merck Index: 14,1567 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...