Ammonium sulfate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

A11682-0B A11682-0I
CAAAAA11682-0BEA 82.63 CAD
CAAAAA11682-0B CAAAAA11682-0I
Ammonium sulfate 98+%
Ammonium sulphate

Purity: 98+% CAS No.: 7783-20-2 Molecular Formula: H8N2O4S Formula Weight: 132.14 Melting Point: ca 280° dec. Merck Reference: 14,555 MDL No.: MFCD00003391

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: ∼ 280 °C (dec.)
Density: 1.769
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 14,555

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR