Lauric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dodecanoic acid, Dodecylic acid

A11672-0C A11672-0E A11672-36
CAAAAA11672-0CEA 360.24 CAD
CAAAAA11672-0C CAAAAA11672-0E CAAAAA11672-36
Lauric acid 98%
Lauric acid

Purity: 98% CAS No.: 143-07-7 Molecular Formula: CH3(CH2)10CO2H Formula Weight: 200.32 Flash Point: 165°(329°F) Odor: Odorless Boiling Point: 180°/21mm Beilstein Reference: 1099477 Merck Reference: 14,5384 MDL No.: MFCD00002736

Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 180 °C (21 mmHg)
Melting Pt: 43…45 °C
Density: 0.883
Flash Pt: 165 °C (329 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 14,5384

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR