Potassium chloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Potassiumchloride

A11662-0B A11662-0G A11662-0I
CAAAAA11662-0BEA 90.85 CAD
CAAAAA11662-0B CAAAAA11662-0G CAAAAA11662-0I
Potassium chloride 99%
Potassium chloride

Purity: 99% CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Refractive Index: 1.490 Odor: Odorless Boiling Point: 1500° subl. Merck Reference: 14,7621 MDL No.: MFCD00011360

Formula: KCl
Boiling Pt: 1500 °C (subl.)
Melting Pt: 773 °C
Density: 1.984
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,7621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR