1-Bromo-4-fluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11630-0E
A11630-22
A11630-36
CAAAAA11630-0EEA
1003.3
CAD
CAAAAA11630-0E
CAAAAA11630-22
CAAAAA11630-36
1-Bromo-4-fluorobenzene 99%
1-Bromo-4-fluorobenzene
Purity: 99%
CAS No.: 460-00-4
Molecular Formula: C6H4BrF
Formula Weight: 175.01
Flash Point: 60°(140°F)
Refractive Index: 1.5270
Odor: Aromatic
Melting Point: -16°
Boiling Point: 151-153°
Beilstein Reference: 774102
UN No.: 1993
MDL No.: MFCD00000342
Formula:
C₆H₄BrF Boiling Pt: 151…153 °C Melting Pt: –16 °C Density: 1.604 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000342 CAS Number: 460-00-4 EINECS: 207-300-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...