2,4-Difluorophenylhydrazinium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A11625-06 A11625-14 A11625-22
CAAAA11625-06EA 90.69 CAD
CAAAA11625-06 CAAAA11625-14 CAAAA11625-22
2,4-Difluorophenylhydrazinium chloride 98%
2,4-Difluorophenylhydrazinium chloride

Purity: 98% CAS No.: 51523-79-6 Molecular Formula: C6H6F2N2.HCl Formula Weight: 180.59 Melting Point: ca 255° dec. Beilstein Reference: 5411448

Formula: C₆H₇ClF₂N₂
Melting Pt: ∼ 255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013383
CAS Number: 51523-79-6
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR