3-Bromopropionic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Bromopropanoic acid
A11573-14
A11573-22
A11573-36
CAAAAA11573-14EA
46.14
CAD
CAAAAA11573-14
CAAAA11573-22
CAAAA11573-36
3-Bromopropionic acid 97%
3-Bromopropionic acid
MDL: MFCD00002763
Beilstein: 1071333
Soluble in water, alcohol, ether, chloroform, benzene
Purity: 97%
CAS No.: 590-92-1
Molecular Formula: BrCH2CH2CO2H
Formula Weight: 152.98
Flash Point: 65°(149°F)
Melting Point: 57-63°
Boiling Point: 140-142°/45mm
Solubility: Soluble in water, alcohol, ether, chloroform, benzene
Beilstein Reference: 1071333
Merck Reference: 14,1433
UN No.: 3261
MDL No.: MFCD00002763
Formula:
BrH₂CCH₂COOH Boiling Pt: 140…142 °C/45 mm Melting Pt: 57…63 °C Density: 1.480 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002763 CAS Number: 590-92-1 EINECS: 209-694-1 UN: 3261 ADR: 8,III Merck Index: 14,1433 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...