Tetrapropylammonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11522-0E A11522-22 A11522-36
CAAAAA11522-0EEA 1482.04 CAD
CAAAAA11522-0E CAAAAA11522-22 CAAAAA11522-36
Tetrapropylammonium bromide 98+%
Tetrapropylammonium bromide

Purity: 98+% CAS No.: 1941-30-6 Molecular Formula: (CH3(CH2)2)4NBr Formula Weight: 266.27 Melting Point: ca 270° dec. Beilstein Reference: 3567846

Formula: C₁₂H₂₈BrN
Melting Pt: ∼ 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011840
CAS Number: 1941-30-6
EINECS: 217-727-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR