1,4,8,11-Tetraazacyclotetradecane 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: CYCLAM

A11516-03 A11516-06 A11516-14
CAAAAA11516-03EA 118.5 CAD
CAAAAA11516-03 CAAAAA11516-06 CAAAAA11516-14
1,4,8,11-Tetraazacyclotetradecane 98+%
1,4,8,11-Tetraazacyclotetradecane

Purity: 98+% CAS No.: 295-37-4 Molecular Formula: C10H24N4 Formula Weight: 200.33 Melting Point: 184-188° Beilstein Reference: 111811

Formula: C₁₀H₂₄N₄
MW: 200.33 g/mol
Melting Pt: 184…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00005105
CAS Number: 295-37-4
EINECS: 206-039-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR