Methyl-3,5-dimethoxybenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 3,5-dimethoxybenzoate, 3,5-Dimethoxybenzoic acid methyl ester

A11486-09 A11486-18
CAAAA11486-09EA 49.77 CAD
CAAAA11486-09 CAAAA11486-18 CAAAA11486-30
Methyl-3,5-dimethoxybenzoate 98%
Methyl 3,5-dimethoxybenzoate

Purity: 98% CAS No.: 2150-37-0 Molecular Formula: C10H12O4 Formula Weight: 196.20 Flash Point: 110°(230°F) Melting Point: 40-43° Boiling Point: 298° Beilstein Reference: 2693126 MDL No.: MFCD00008432

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 298 °C
Melting Pt: 40…43 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008432
CAS Number: 2150-37-0
EINECS: 218-423-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR