Carbazole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11448-18
A11448-22
A11448-36
CAAAAA11448-18EA
58.14
CAD
CAAAAA11448-18
CAAAAA11448-22
CAAAAA11448-36
Carbazole 95%
Carbazole
Purity: 95%
CAS No.: 86-74-8
Molecular Formula: C12H9N
Formula Weight: 167.21
Flash Point: 220°(428°F)
Odor: Odorless
Boiling Point: 354-356°
Beilstein Reference: 3956
Merck Reference: 14,1790
UN No.: 3077
MDL No.: MFCD00004960
Formula:
C₁₂H₉N Boiling Pt: 354…356 °C Melting Pt: 240…246 °C Density: 1.150 Flash Pt: 220 °C (428 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004960 CAS Number: 86-74-8 EINECS: 201-696-0 UN: 3077 ADR: 9,III Merck Index: 14,1790 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...